Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004577.3 | NP_004568.2 | 7 | 1177 | ||
XM_005271773.1 | XP_005271830.1 | 7 | 1247 | |||
XM_005271774.1 | XP_005271831.1 | 8 | 1363 | |||
XM_005271775.1 | XP_005271832.1 | 7 | 1203 | |||
XM_005271776.1 | XP_005271833.1 | 8 | 1311 | |||
XM_006715760.1 | XP_006715823.1 | 8 | 1361 | |||
XM_006715761.1 | XP_006715824.1 | 6 | 1112 | |||
XM_011515461.1 | XP_011513763.1 | 8 | 1317 | |||
XM_017012466.1 | XP_016867955.1 | 9 | 1477 | |||
XM_017012467.1 | XP_016867956.1 | 9 | 1573 | |||
XM_017012468.1 | XP_016867957.1 | 7 | 1226 | |||
XM_017012469.1 | XP_016867958.1 | 7 | 1176 | |||
GenBank mRNA | AK223508.1 | - | 4 | 467 | ||
AK315235.1 | - | 6 | 789 | |||
BC063614.1 | - | 7 | 1025 | 3850060 | ||
BX537439.1 | - | 7 | 1147 |