Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000264.3 | NP_000255.2 | 7 | 1216 | ||
NM_001083602.1 | NP_001077071.1 | 7 | 1324 | |||
NM_001083603.1 | NP_001077072.1 | 7 | 1170 | |||
NM_001083604.1 | NP_001077073.1 | 7 | 934 | |||
NM_001083605.1 | NP_001077074.1 | 7 | 917 | |||
NM_001083606.1 | NP_001077075.1 | 7 | 1016 | |||
NM_001083607.1 | NP_001077076.1 | 7 | 1090 | |||
GenBank mRNA | AB209495.1 | - | 4 | 665 | ||
AB233422.1 | - | 2 | 168 | |||
U43148.1 | - | 7 | 1016 | |||
U59464.1 | - | 7 | 1028 |