Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000954.5 | NP_000945.3 | 5 | 557 | ||
GenBank mRNA | AA621632.1 | - | 3 | 267 | ||
AK075333.1 | - | 5 | 557 | |||
AK312817.1 | - | 5 | 544 | |||
AY026356.1 | - | 5 | 482 | |||
BC005939.1 | - | 5 | 491 | 4294999 | ||
BC041463.1 | - | 7 | 1765 | |||
BM686612.1 | - | 5 | 545 | |||
BT019921.1 | - | 5 | 482 | |||
BT019922.1 | - | 5 | 482 | |||
M61900.2 | - | 5 | 547 |