Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000956.3 | NP_000947.2 | 2 | 1119 | ||
GenBank mRNA | AY275471.1 | - | 2 | 868 | ||
BC093927.1 | - | 2 | 921 | 7939772 | ||
BC093929.1 | - | 2 | 921 | 7939774 | ||
BE739744.1 | - | 1 | 666 | |||
U19487.1 | - | 2 | 1024 | |||
X83868.1 | - | 2 | 868 |
Set Membership: |
G-protein coupled receptor |