Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002836.3 | NP_002827.1 | 28 | 3065 | ||
NM_080840.2 | NP_543030.1 | 23 | 2756 | |||
NM_080841.2 | NP_543031.1 | 23 | 2581 | |||
GenBank mRNA | AB209702.1 | - | 23 | 3911 | ||
AK090743.1 | - | 18 | 2169 | |||
AK290233.1 | - | 23 | 2695 | |||
AK294499.1 | - | 23 | 2667 | |||
AK297293.1 | - | 18 | 1981 | |||
AK297647.1 | - | 18 | 1956 | |||
BC027308.1 | - | 23 | 2561 | 3920094 | ||
BX571753.1 | - | 24 | 2807 | |||
M34668.1 | - | 29 | 3085 | |||
X53364.1 | - | 23 | 2541 | |||
X54130.1 | - | 21 | 2393 | |||
X54890.1 | - | 23 | 2549 |