Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002851.2 | NP_002842.2 | 12 | 5215 | ||
XM_005250519.1 | XP_005250576.1 | 12 | 5215 | |||
XM_017012477.1 | XP_016867966.1 | 13 | 5262 | |||
GenBank mRNA | M93426.1 | - | 12 | 4964 | ||
X54135.1 | - | 1 | 383 |