Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206838.1 | NP_001193767.1 | 13 | 2718 | ||
NM_001206839.1 | NP_001193768.1 | 13 | 2718 | |||
NM_002851.2 | NP_002842.2 | 13 | 5298 | |||
XM_005250519.1 | XP_005250576.1 | 13 | 5298 | |||
XM_017012477.1 | XP_016867966.1 | 14 | 5345 | |||
GenBank mRNA | AK294062.1 | - | 2 | 552 | ||
M93426.1 | - | 13 | 5047 | |||
X54135.1 | - | 2 | 466 |