Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002853.3 | NP_002844.1 | 4 | 714 | ||
NR_026591.1 | 3 | 556 | ||||
GenBank mRNA | AF011905.1 | - | 4 | 599 | ||
AF030933.1 | - | 6 | 822 | |||
AF058392.1 | - | 3 | 385 | |||
AF058393.1 | - | 2 | 276 | |||
AF073524.1 | - | 4 | 580 | |||
AF074717.1 | - | 4 | 524 | |||
AF076841.1 | - | 4 | 580 | |||
AF084512.1 | - | 4 | 589 | |||
AF084513.1 | - | 3 | 480 | |||
AF090170.1 | - | 3 | 471 | |||
AJ004974.1 | - | 4 | 548 | |||
AJ004975.1 | - | 3 | 439 | |||
AK002112.1 | - | 4 | 717 | |||
BC006837.1 | - | 4 | 566 | 3450321 | ||
BC009804.1 | - | 4 | 541 | 4109436 | ||
BC037857.1 | - | 4 | 1813 | 5272408 | ||
BT006908.1 | - | 3 | 385 |