Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PURA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005859.4 | NP_005850.1 | 1 | 775 | ||
GenBank mRNA | BC036087.1 | - | 1 | 728 | 5284976 | |
BC110574.1 | - | 2 | 605 | |||
BC110594.1 | - | 2 | 654 | |||
BT019388.1 | - | 1 | 716 | |||
M96684.1 | - | 1 | 775 |