Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PURB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033224.4 | NP_150093.1 | 1 | 811 | ||
GenBank mRNA | AY039216.1 | - | 1 | 787 | ||
BC101735.1 | - | 1 | 793 | 8069241 | ||
BC101737.1 | - | 1 | 793 | 8069243 | ||
BU177276.1 | - | 1 | 211 |