Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135768.2 | NP_001129240.1 | 8 | 1372 | ||
NM_001135769.2 | NP_001129241.1 | 7 | 1348 | |||
NM_006505.4 | NP_006496.4 | 8 | 1507 | |||
GenBank mRNA | AK094177.1 | - | 8 | 1314 | ||
AK290946.1 | - | 8 | 1421 | |||
AK300349.1 | - | 8 | 1260 | |||
BC015542.1 | - | 8 | 1379 | 3902226 | ||
DA849917.1 | - | 3 | 140 | |||
M24407.1 | - | 8 | 1208 |