Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135768.2 | NP_001129240.1 | 4 | 1123 | ||
NM_001135769.2 | NP_001129241.1 | 4 | 1123 | |||
NM_001135770.3 | NP_001129242.2 | 4 | 1123 | |||
NM_006505.4 | NP_006496.4 | 4 | 1123 | |||
GenBank mRNA | AK094177.1 | - | 4 | 930 | ||
AK290946.1 | - | 4 | 1037 | |||
AK300349.1 | - | 4 | 1011 | |||
BC015542.1 | - | 4 | 995 | 3902226 | ||
M24406.1 | - | 4 | 824 | |||
M24407.1 | - | 4 | 824 |