Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECTIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042724.1 | NP_001036189.1 | 2, 3 | 858 | ||
NM_002856.2 | NP_002847.1 | 2, 3 | 858 | |||
XM_011527192.2 | XP_011525494.1 | 2, 3 | 858 | |||
GenBank mRNA | AF058448.1 | - | 2, 3 | 617 | ||
AK291330.1 | - | 2, 3 | 684 | |||
BC003091.1 | - | 2, 3 | 666 | 3503222 | ||
CR456818.1 | - | 2, 3 | 468 | |||
X80038.1 | - | 2, 3 | 468 |