Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PXN SIRNA Assays ›
Gene Symbol:
PXN
Gene Name:
paxillin
Gene Aliases:
-
Chromosome Location:
Chr.12: 120210439 - 120265771 on Build GRCh38
UniGene ID:
Hs.446336
Species:
Human
siRNA ID
HSS108926
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PXN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080855.2
NP_001074324.1
5
743
NM_001243756.1
NP_001230685.1
5
743
NM_002859.3
NP_002850.2
5
743
NM_025157.4
NP_079433.3
5
867
XM_006719532.1
XP_006719595.1
5
712
XM_006719534.1
XP_006719597.1
5
867
XM_006719535.3
XP_006719598.1
6
795
XM_006719536.2
XP_006719599.1
5
684
XM_011538622.1
XP_011536924.1
6
794
XM_011538623.1
XP_011536925.1
6
981
XM_017019726.1
XP_016875215.1
5
1280
XM_017019727.1
XP_016875216.1
5
1080
XM_017019728.1
XP_016875217.1
5
1071
XM_017019729.1
XP_016875218.1
5
1280
XM_017019730.1
XP_016875219.1
5
1280
XM_017019731.1
XP_016875220.1
6
1165
XM_017019732.1
XP_016875221.1
5
1280
XM_017019733.1
XP_016875222.1
5
1280
XM_017019735.1
XP_016875224.1
5
1280
XM_017019736.1
XP_016875225.1
5
1280
XM_017019737.1
XP_016875226.1
5
1280
XM_017019738.1
XP_016875227.1
5
1280
XM_017019739.1
XP_016875228.1
5
1280
XM_017019740.1
XP_016875229.1
5
1280
XM_017019741.1
XP_016875230.1
5
1071
XM_017019742.1
XP_016875231.1
5
1280
XM_017019743.1
XP_016875232.1
6
981
GenBank mRNA
AB209034.1
-
5
690
AK122964.1
-
5
1071
AK314204.1
-
5
702
BC136787.1
-
5
677
9020777
BC136794.1
-
5
677
9020784
BC144410.1
-
5
677
9052931
BU500419.1
-
5
676
BX648777.1
-
5
861
D86862.1
-
4
601
D86863.1
-
4
601
U14588.1
-
5
675
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell adhesion
cell-matrix adhesion
signal transduction
signal complex assembly
epidermal growth factor receptor signaling pathway
transforming growth factor beta receptor signaling pathway
cellular response to reactive oxygen species
vascular endothelial growth factor receptor signaling pathway
growth hormone receptor signaling pathway
integrin binding
protein binding
beta-catenin binding
zinc ion binding
vinculin binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline