Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002862.3 | NP_002853.2 | 7 | 949 | ||
GenBank mRNA | AB209080.1 | - | 7 | 907 | ||
AK299697.1 | - | 7 | 873 | |||
AK309036.1 | - | 7 | 947 | |||
BC017045.2 | - | 7 | 924 | 3877951 | ||
BC030795.2 | - | 7 | 925 | 3916938 | ||
J03544.1 | - | 7 | 919 | |||
U47025.1 | - | 7 | 874 |