Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272073.1 | NP_001259002.1 | 23 | 2530 | ||
NM_005051.2 | NP_005042.1 | 23 | 2563 | |||
NR_073590.1 | 23 | 2514 | ||||
GenBank mRNA | AF130067.1 | - | 19 | 1691 | ||
AK056815.1 | - | 23 | 2142 | |||
AK222510.1 | - | 23 | 2191 | |||
AK290523.1 | - | 23 | 2191 | |||
AK293260.1 | - | 22 | 2090 | |||
AK297987.1 | - | 22 | 2037 | |||
AK301559.1 | - | 23 | 2159 | |||
BC000394.2 | - | 23 | 2172 | 2821788 | ||
BC001567.1 | - | 23 | 2170 | 3452261 | ||
BC001772.1 | - | 19 | 1698 | |||
BC016634.1 | - | 22 | 1995 | |||
BC029739.1 | - | 23 | 2173 | 3857444 | ||
X76013.1 | - | 23 | 2172 |