Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272073.1 | NP_001259002.1 | 10, 11 | 1245 | ||
NM_005051.2 | NP_005042.1 | 10, 11 | 1278 | |||
NR_073590.1 | 10, 11 | 1229 | ||||
XM_017006965.1 | XP_016862454.1 | 10, 11 | 958 | |||
GenBank mRNA | AF130067.1 | - | 6, 7 | 406 | ||
AK056815.1 | - | 10, 11 | 857 | |||
AK222510.1 | - | 10, 11 | 906 | |||
AK290523.1 | - | 10, 11 | 906 | |||
AK293260.1 | - | 9, 10 | 805 | |||
AK297987.1 | - | 9, 10 | 752 | |||
AK300220.1 | - | 9, 10 | 843 | |||
AK301559.1 | - | 10, 11 | 874 | |||
BC000394.2 | - | 10, 11 | 887 | 2821788 | ||
BC001567.1 | - | 10, 11 | 885 | 3452261 | ||
BC001772.1 | - | 6, 7 | 413 | |||
BC016634.1 | - | 9, 10 | 710 | |||
BC029739.1 | - | 10, 11 | 888 | 3857444 | ||
X76013.1 | - | 10, 11 | 887 |