Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QDPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000320.2 | NP_000311.2 | 7 | 860 | ||
NM_001306140.1 | NP_001293069.1 | 6 | 767 | |||
GenBank mRNA | AK124382.1 | - | 6 | 619 | ||
AK223437.1 | - | 7 | 716 | |||
AK289773.1 | - | 7 | 716 | |||
BC000576.2 | - | 7 | 704 | 3162690 | ||
M16447.1 | - | 7 | 760 | |||
X04882.1 | - | 7 | 704 |