Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242644.1 | NP_001229573.1 | 3, 4 | 501 | ||
NM_002865.2 | NP_002856.1 | 4, 5 | 573 | |||
GenBank mRNA | AF498930.1 | - | 4, 5 | 275 | ||
AK297576.1 | - | 3, 4 | 475 | |||
AK312344.1 | - | 4, 5 | 496 | |||
BC008929.2 | - | 4, 5 | 463 | 2966694 | ||
BT019695.1 | - | 4, 5 | 275 | |||
M28213.1 | - | 4, 5 | 315 | |||
X12953.1 | - | 4, 5 | 483 | |||
X13899.1 | - | 4, 5 | 275 |