Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002866.4 | NP_002857.1 | 5 | 732 | ||
XM_011528164.1 | XP_011526466.1 | 5 | 729 | |||
GenBank mRNA | AF254795.1 | - | 5 | 692 | ||
AF498931.1 | - | 4 | 563 | |||
AK289559.1 | - | 5 | 710 | |||
AK293467.1 | - | 922 | ||||
BC011782.2 | - | 5 | 697 | 3939176 | ||
BM717329.1 | - | 1 | 26 | |||
M28210.1 | - | 5 | 602 | |||
X13900.1 | - | 5 | 602 |