Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292048.1 | NP_001278977.1 | 2 | 624 | ||
NM_004162.4 | NP_004153.2 | 2 | 624 | |||
GenBank mRNA | AF464088.1 | - | 2 | 465 | ||
AF498936.1 | - | 1 | 89 | |||
AK295992.1 | - | 2 | 299 | |||
AK309287.1 | - | 2 | 305 | |||
AK312618.1 | - | 2 | 338 | |||
BC001267.1 | - | 2 | 254 | 3463669 | ||
BC018288.1 | - | 2 | 307 | 3454268 | ||
CR536492.1 | - | 1 | 89 | |||
DA972638.1 | - | 2 | 413 | |||
M28215.1 | - | 1 | 145 | |||
X13903.1 | - | 1 | 89 |