Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002871.4 | NP_002862.2 | 2 | 486 | ||
GenBank mRNA | AK223538.1 | - | 2 | 486 | ||
AK311900.1 | - | 2 | 377 | |||
BC018488.1 | - | 2 | 339 | 4475846 | ||
BC037392.1 | - | 2 | 364 | 5191960 | ||
BT007133.1 | - | 2 | 327 | |||
S78873.1 | - | 2 | 339 | |||
U74324.1 | - | 2 | 330 |