Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002881.2 | NP_002872.1 | 3 | 337 | ||
XM_005263724.1 | XP_005263781.1 | 3 | 250 | |||
XM_005263727.1 | XP_005263784.1 | 3 | 339 | |||
XM_005263728.1 | XP_005263785.1 | 3 | 544 | |||
XM_005263729.2 | XP_005263786.1 | 3 | 595 | |||
XM_011511573.1 | XP_011509875.1 | 3 | 247 | |||
XM_011511574.1 | XP_011509876.1 | 4 | 398 | |||
XM_017004621.1 | XP_016860110.1 | 3 | 288 | |||
XM_017004622.1 | XP_016860111.1 | 4 | 436 | |||
GenBank mRNA | AF493911.1 | - | 2 | 147 | ||
AK127675.1 | - | 4 | 385 | |||
AK303214.1 | - | 3 | 256 | |||
AK312453.1 | - | 2 | 161 | |||
BC018163.1 | - | 3 | 242 | 3880116 | ||
BQ893549.1 | - | 342 | ||||
BT006953.1 | - | 2 | 147 | |||
M35416.1 | - | 3 | 317 | |||
X15015.1 | - | 2 | 147 |