Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278651.1 | NP_001265580.1 | 16 | 2403 | ||
NM_001317930.1 | NP_001304859.1 | 15 | 1857 | |||
NM_002883.3 | NP_002874.1 | 15 | 1942 | |||
XM_005261695.1 | XP_005261752.1 | 15 | 2070 | |||
XM_005261696.1 | XP_005261753.1 | 15 | 1841 | |||
XM_006724289.3 | XP_006724352.1 | 15 | 1845 | |||
XM_011530293.1 | XP_011528595.1 | 14 | 1935 | |||
XM_011530294.2 | XP_011528596.1 | 15 | 3551 | |||
XM_011530295.2 | XP_011528597.1 | 15 | 2250 | |||
XM_017028893.1 | XP_016884382.1 | 16 | 3181 | |||
XM_017028894.1 | XP_016884383.1 | 16 | 3319 | |||
XM_017028895.1 | XP_016884384.1 | 15 | 3681 | |||
XM_017028896.1 | XP_016884385.1 | 16 | 2069 | |||
XM_017028897.1 | XP_016884386.1 | 15 | 3161 | |||
XM_017028898.1 | XP_016884387.1 | 17 | 3094 | |||
XM_017028899.1 | XP_016884388.1 | 14 | 1807 | |||
GenBank mRNA | AB058738.1 | - | 14 | 3126 | ||
AK302222.1 | - | 14 | 1506 | |||
AK303524.1 | - | 15 | 1865 | |||
BC004990.1 | - | 5 | 517 | |||
BC014044.2 | - | 15 | 1857 | 3506039 | ||
BC019676.1 | - | 1 | 58 | |||
BC041396.2 | - | 16 | 2413 | 5277434 | ||
CR456557.1 | - | 14 | 1699 | |||
X82260.1 | - | 14 | 1638 |