Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAP1GDS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100426.1 | NP_001093896.1 | 9 | 1162 | ||
NM_001100427.1 | NP_001093897.1 | 9 | 1159 | |||
NM_001100428.1 | NP_001093898.1 | 8 | 1015 | |||
NM_001100429.1 | NP_001093899.1 | 8 | 1012 | |||
NM_001100430.1 | NP_001093900.1 | 7 | 886 | |||
NM_021159.4 | NP_066982.3 | 9 | 1162 | |||
GenBank mRNA | AF215923.1 | - | 8 | 969 | ||
AF237413.1 | - | 8 | 969 | |||
AK055032.1 | - | 9 | 1082 | |||
AK295156.1 | - | 8 | 974 | |||
AK297543.1 | - | 6 | 709 | |||
AK311243.1 | - | 6 | 697 | |||
AY376900.1 | - | 6 | 696 | |||
BC001816.2 | - | 9 | 1069 | 3010166 | ||
BC001851.2 | - | 9 | 1069 | 3010166 | ||
BC098269.1 | - | 9 | 1020 | 40001059 | ||
BC098334.1 | - | 8 | 870 | 40001057 | ||
BC099708.1 | - | 8 | 870 | 40001058 | ||
BC099845.1 | - | 8 | 873 | 40001061 | ||
BT006837.1 | - | 8 | 972 | |||
X63465.1 | - | 7 | 822 |