Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000966.5 | NP_000957.1 | 10 | 1722 | ||
NM_001042728.2 | NP_001036193.1 | 8 | 1510 | |||
NM_001243730.1 | NP_001230659.1 | 9 | 1396 | |||
NM_001243731.1 | NP_001230660.1 | 8 | 1247 | |||
NM_001243732.1 | NP_001230661.1 | 7 | 1361 | |||
GenBank mRNA | AB307689.1 | - | 8 | 1230 | ||
AK290588.1 | - | 10 | 1718 | |||
AK297023.1 | - | 9 | 1314 | |||
AK297110.1 | - | 8 | 1223 | |||
AK297277.1 | - | 7 | 1361 | |||
AK316259.1 | - | 8 | 1234 | |||
BC093727.1 | - | 8 | 1254 | 7939572 | ||
BC093729.1 | - | 8 | 1254 | 7939574 | ||
HQ692828.1 | - | 8 | 1230 | |||
HQ692829.1 | - | 8 | 1197 | |||
L12060.1 | - | 9 | 1338 | |||
M24857.1 | - | 10 | 1644 | |||
M38258.1 | - | 8 | 1376 | |||
M57707.1 | - | 9 | 1429 |