Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000966.5 | NP_000957.1 | 5, 6 | 974 | ||
NM_001042728.2 | NP_001036193.1 | 3, 4 | 762 | |||
NM_001243730.1 | NP_001230659.1 | 4, 5 | 648 | |||
NM_001243731.1 | NP_001230660.1 | 3, 4 | 499 | |||
NM_001243732.1 | NP_001230661.1 | 2, 3 | 613 | |||
GenBank mRNA | AB307689.1 | - | 3, 4 | 482 | ||
AK290588.1 | - | 5, 6 | 970 | |||
AK297023.1 | - | 4, 5 | 566 | |||
AK297110.1 | - | 3, 4 | 475 | |||
AK297277.1 | - | 2, 3 | 613 | |||
AK297801.1 | - | 3, 4 | 608 | |||
AK301057.1 | - | 3, 4 | 757 | |||
AK316259.1 | - | 3, 4 | 486 | |||
BC093727.1 | - | 3, 4 | 506 | 7939572 | ||
BC093729.1 | - | 3, 4 | 506 | 7939574 | ||
HQ692828.1 | - | 3, 4 | 482 | |||
HQ692829.1 | - | 3, 4 | 449 | |||
L12060.1 | - | 4, 5 | 590 | |||
M24857.1 | - | 5, 6 | 896 | |||
M38258.1 | - | 3, 4 | 628 | |||
M57707.1 | - | 4, 5 | 681 |