Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARRES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002888.3 | NP_002879.2 | 4 | 664 | ||
NM_206963.1 | NP_996846.1 | 4 | 664 | |||
XM_005247686.4 | XP_005247743.3 | 4 | 1080 | |||
GenBank mRNA | AK130079.1 | - | 4 | 635 | ||
AW514087.1 | - | 1 | 219 | |||
BC029640.1 | - | 4 | 600 | 4776506 | ||
BM919188.1 | - | 4 | 791 | |||
U27185.1 | - | 4 | 629 |