Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARRES3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004585.4 | NP_004576.2 | 4 | 512 | ||
GenBank mRNA | AB030815.1 | - | 4 | 468 | ||
AB453252.1 | - | 3 | 444 | |||
AF060228.1 | - | 4 | 473 | |||
AF092922.1 | - | 4 | 505 | |||
AK312110.1 | - | 4 | 497 | |||
BC009678.1 | - | 4 | 492 | 3877277 | ||
DA973023.1 | - | 4 | 514 |