Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145648.1 | NP_001139120.1 | 17 | 3000 | ||
NM_002891.4 | NP_002882.3 | 18 | 3048 | |||
NM_153815.2 | NP_722522.1 | 4 | 1494 | |||
XM_011521866.2 | XP_011520168.1 | 18 | 2948 | |||
XM_011521867.2 | XP_011520169.1 | 18 | 2948 | |||
XM_017022455.1 | XP_016877944.1 | 18 | 2939 | |||
XM_017022456.1 | XP_016877945.1 | 17 | 2909 | |||
XM_017022457.1 | XP_016877946.1 | 17 | 2909 | |||
XM_017022458.1 | XP_016877947.1 | 17 | 2900 | |||
XM_017022459.1 | XP_016877948.1 | 17 | 2909 | |||
GenBank mRNA | AK226101.1 | - | 16 | 2284 | ||
AK301205.1 | - | 6 | 1097 | |||
AK309047.1 | - | 16 | 2431 | |||
BC040275.1 | - | 17 | 2891 | 5192914 | ||
L26584.1 | - | 18 | 2679 | |||
S62035.1 | - | 4 | 1503 |