Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145648.1 | NP_001139120.1 | 26 | 4024 | ||
NM_002891.4 | NP_002882.3 | 27 | 4072 | |||
NM_153815.2 | NP_722522.1 | 13 | 2518 | |||
XM_011521867.2 | XP_011520169.1 | 26 | 3854 | |||
XM_017022455.1 | XP_016877944.1 | 27 | 3963 | |||
XM_017022456.1 | XP_016877945.1 | 26 | 3933 | |||
XM_017022459.1 | XP_016877948.1 | 25 | 3815 | |||
GenBank mRNA | AK226101.1 | - | 25 | 3308 | ||
L26584.1 | - | 27 | 3703 | |||
M91815.1 | - | 6 | 565 | |||
S62035.1 | - | 13 | 2527 |