Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002897.4 | NP_002888.1 | 12, 13 | 1579 | ||
NM_016836.3 | NP_058520.1 | 12, 13 | 1588 | |||
XM_005246737.1 | XP_005246794.1 | 13, 14 | 1347 | |||
XM_005246738.2 | XP_005246795.1 | 13, 14 | 1682 | |||
XM_005246739.2 | XP_005246796.1 | 13, 14 | 1552 | |||
XM_005246740.1 | XP_005246797.1 | 13, 14 | 1338 | |||
XM_005246741.3 | XP_005246798.1 | 13, 14 | 1549 | |||
XM_017004626.1 | XP_016860115.1 | 12, 13 | 1625 | |||
GenBank mRNA | AK127157.1 | - | 3, 4 | 1980 | ||
AK225887.1 | - | 13, 14 | 1247 | |||
AK297812.1 | - | 13, 14 | 1338 | |||
BC012992.2 | - | 12, 13 | 1274 | 3530897 | ||
BC012993.2 | - | 12, 13 | 1274 | 3530898 | ||
BC018951.2 | - | 12, 13 | 1575 | 4137722 | ||
BC051889.1 | - | 12, 13 | 1161 | |||
BX648246.1 | - | 5, 6 | 8745 | |||
D28482.1 | - | 12, 13 | 1145 | |||
KU178305.1 | - | 12, 13 | 1135 | |||
KU178306.1 | - | 13, 14 | 1189 | |||
L11289.1 | - | 12, 13 | 1212 | |||
X64652.1 | - | 11, 12 | 1094 | |||
X77494.1 | - | 14, 15 | 1324 |