Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130992.1 | NP_001124464.1 | 2 | 520 | ||
NM_001130993.1 | NP_001124465.1 | 2 | 520 | |||
NM_002899.3 | NP_002890.2 | 2 | 520 | |||
GenBank mRNA | AK290315.1 | - | 2 | 327 | ||
AK301684.1 | - | 2 | 410 | |||
AK307378.1 | - | 2 | 329 | |||
AK309492.1 | - | 2 | 520 | |||
BC121052.2 | - | 2 | 278 | 40118103 | ||
BU734286.1 | - | 3 | 393 | |||
CR541979.1 | - | 2 | 223 | |||
CR542005.1 | - | 2 | 223 | |||
DC394326.1 | - | 2 | 410 | |||
M11433.1 | - | 2 | 348 | |||
M36809.1 | - | 2 | 295 |