Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004164.2 | NP_004155.2 | 2 | 288 | ||
GenBank mRNA | AI076320.1 | - | 3 | 409 | ||
AK291978.1 | - | 2 | 294 | |||
AW772327.1 | - | 3 | 397 | |||
BC069296.1 | - | 2 | 237 | 7262683 | ||
BC069361.1 | - | 2 | 237 | 7262695 | ||
BC069396.1 | - | 2 | 237 | 7262707 | ||
BC069424.1 | - | 2 | 237 | 7262719 | ||
BC069447.1 | - | 2 | 237 | 7262731 | ||
BC069469.1 | - | 2 | 236 | |||
BC069513.1 | - | 2 | 237 | 7262755 | ||
BC069522.1 | - | 2 | 237 | 7262672 | ||
BX282005.1 | - | 4 | 415 |