Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002901.2 | NP_002892.1 | 5 | 1085 | ||
GenBank mRNA | AK094360.1 | - | 4 | 1175 | ||
AK126419.1 | - | 1 | 1416 | |||
AK129791.1 | - | 5 | 1112 | |||
AY423728.1 | - | 3 | 321 | |||
BC010120.2 | - | 5 | 954 | 3836263 | ||
D42073.1 | - | 5 | 871 | |||
FJ224346.1 | - | 5 | 871 |