Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002902.2 | NP_002893.1 | 3, 4 | 674 | ||
GenBank mRNA | AK313833.1 | - | 3, 4 | 545 | ||
AL110102.1 | - | 2, 3 | 251 | |||
BC004892.2 | - | 3, 4 | 514 | 3505241 | ||
BT009790.1 | - | 3, 4 | 453 | |||
X78669.1 | - | 3, 4 | 519 |