Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000537.3 | NP_000528.1 | 2 | 237 | ||
GenBank mRNA | AF117822.1 | - | 2 | 166 | ||
BC033474.1 | - | 2 | 219 | 5188566 | ||
BC047752.1 | - | 2 | 222 | 5590211 | ||
BP321283.1 | - | 2 | 237 |