Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278791.1 | NP_001265720.1 | 7 | 780 | ||
NM_001278792.1 | NP_001265721.1 | 6 | 571 | |||
NM_001278793.1 | NP_001265722.1 | 6 | 678 | |||
NM_002914.4 | NP_002905.2 | 6 | 576 | |||
NM_181471.2 | NP_852136.1 | 7 | 678 | |||
XM_006716080.2 | XP_006716143.1 | 5 | 400 | |||
XM_017012492.1 | XP_016867981.1 | 7 | 656 | |||
XM_017012493.1 | XP_016867982.1 | 5 | 424 | |||
GenBank mRNA | AB451243.1 | - | 7 | 603 | ||
AB451368.1 | - | 7 | 603 | |||
BC002813.1 | - | 6 | 506 | 3637774 | ||
BM563468.1 | - | 7 | 709 | |||
BM810551.1 | - | 6 | 483 | |||
BQ070526.1 | - | 7 | 628 | |||
BQ674131.1 | - | 6 | 529 | |||
BT007058.1 | - | 6 | 501 | |||
M87338.1 | - | 810 |