Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002916.3 | NP_002907.1 | 9 | 1059 | ||
NM_181573.2 | NP_853551.1 | 9 | 1118 | |||
GenBank mRNA | AA521171.1 | - | 3 | 365 | ||
BC017452.1 | - | 9 | 996 | 3537752 | ||
BC024022.1 | - | 9 | 1012 | 4658221 | ||
BT006987.1 | - | 8 | 836 | |||
CR536561.1 | - | 8 | 836 |