Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002918.4 | NP_002909.4 | 14 | 2143 | ||
XM_011528165.2 | XP_011526467.1 | 15 | 2473 | |||
XM_011528166.2 | XP_011526468.1 | 15 | 2409 | |||
XM_011528167.2 | XP_011526469.1 | 15 | 3018 | |||
XM_011528168.2 | XP_011526470.1 | 15 | 2179 | |||
XM_011528169.2 | XP_011526471.1 | 14 | 2069 | |||
XM_011528170.2 | XP_011526472.1 | 14 | 2647 | |||
GenBank mRNA | A20498.1 | - | 14 | 1964 | ||
BC049826.1 | - | 14 | 2104 | 6043247 | ||
X58964.1 | - | 14 | 1964 |