Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000449.3 | NP_000440.1 | 11 | 1257 | ||
NM_001025603.1 | NP_001020774.1 | 11 | 1250 | |||
XM_005245405.1 | XP_005245462.1 | 11 | 1312 | |||
XM_005245406.3 | XP_005245463.1 | 11 | 1269 | |||
XM_011509847.1 | XP_011508149.1 | 11 | 1316 | |||
XM_011509848.1 | XP_011508150.1 | 11 | 1319 | |||
XM_011509849.1 | XP_011508151.1 | 11 | 1252 | |||
XM_011509850.1 | XP_011508152.1 | 11 | 1379 | |||
XM_017001999.1 | XP_016857488.1 | 11 | 1380 | |||
XM_017002000.1 | XP_016857489.1 | 11 | 1411 | |||
GenBank mRNA | AK223526.1 | - | 9 | 1049 | ||
AK302891.1 | - | 10 | 1130 | |||
AK312289.1 | - | 11 | 1230 | |||
AL050135.1 | - | 3 | 492 | |||
BC017471.1 | - | 11 | 1184 | 3909092 | ||
X85786.1 | - | 10 | 1197 |