Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102445.2 | NP_001095915.1 | 6 | 1127 | ||
NM_001113380.1 | NP_001106851.1 | 5 | 702 | |||
NM_001113381.1 | NP_001106852.1 | 4 | 571 | |||
NM_005613.5 | NP_005604.1 | 5 | 1018 | |||
GenBank mRNA | AB209019.1 | - | 1 | 3094 | ||
AF493928.1 | - | 5 | 507 | |||
AK093959.1 | - | 4 | 464 | |||
AK096204.1 | - | 3 | 1632 | |||
AK295540.1 | - | 6 | 862 | |||
AK296240.1 | - | 5 | 546 | |||
AK300832.1 | - | 5 | 506 | |||
AK312250.1 | - | 5 | 630 | |||
BC000737.1 | - | 5 | 592 | 3510260 | ||
BC051869.1 | - | 5 | 604 | 5786695 | ||
BT007025.1 | - | 5 | 507 | |||
DQ346663.1 | - | 5 | 846 | |||
DQ346664.1 | - | 6 | 958 | |||
EF054877.1 | - | 5 | 702 | |||
U27768.1 | - | 5 | 604 |