Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RGS7 SIRNA Assays ›
Gene Symbol:
RGS7
Gene Name:
regulator of G-protein signaling 7
Gene Aliases:
-
Chromosome Location:
Chr.1: 240775193 - 241357374 on Build GRCh38
UniGene ID:
Hs.655739
Species:
Human
siRNA ID
HSS109230
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RGS7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282773.1
NP_001269702.1
12
1261
NM_001282775.1
NP_001269704.1
14
1420
NM_001282778.1
NP_001269707.1
14
1420
NM_002924.5
NP_002915.3
14
1420
XM_005273218.3
XP_005273275.1
14
1564
XM_006711800.3
XP_006711863.1
13
1355
XM_011544246.2
XP_011542548.1
13
1014
XM_011544247.2
XP_011542549.1
13
1059
XM_017002001.1
XP_016857490.1
13
1513
XM_017002002.1
XP_016857491.1
15
1439
XM_017002003.1
XP_016857492.1
13
1513
XM_017002004.1
XP_016857493.1
13
1513
XM_017002005.1
XP_016857494.1
14
2455
XM_017002006.1
XP_016857495.1
15
1485
XM_017002007.1
XP_016857496.1
15
1416
XM_017002008.1
XP_016857497.1
15
1444
XM_017002009.1
XP_016857498.1
13
1344
XM_017002010.1
XP_016857499.1
14
1129
XM_017002011.1
XP_016857500.1
11
2526
XM_017002012.1
XP_016857501.1
11
2524
XM_017002013.1
XP_016857502.1
11
2523
GenBank mRNA
AF090116.1
-
14
1275
AF090117.1
-
7
531
AF493930.1
-
11
879
AF493931.1
-
13
1038
AK294234.1
-
14
1254
AK294354.1
-
14
1395
AK314862.1
-
14
1281
AY587875.1
-
13
1038
BC022009.1
-
14
1373
4823760
U32439.1
-
12
892
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
G-protein coupled receptor signaling pathway
negative regulation of signal transduction
intracellular signal transduction
positive regulation of GTPase activity
signal transducer activity
GTPase activator activity
G-protein beta-subunit binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-ffltp:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline