Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000326.4 | NP_000317.1 | 9 | 1320 | ||
XM_011521870.2 | XP_011520172.1 | 9 | 2749 | |||
XM_017022460.1 | XP_016877949.1 | 6 | 1159 | |||
GenBank mRNA | AK312457.1 | - | 9 | 1208 | ||
BC004199.2 | - | 9 | 1222 | 3604048 | ||
BF940774.1 | - | 1 | 414 | |||
BG205561.1 | - | 3 | 415 | |||
BM704704.1 | - | 3 | 327 | |||
J04213.1 | - | 8 | 1123 |