Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002934.2 | NP_002925.1 | 2 | 218 | ||
GenBank mRNA | BC093678.1 | - | 2 | 216 | 7939523 | |
BC093680.1 | - | 2 | 216 | 7939525 | ||
BC096059.3 | - | 2 | 186 | 40000108 | ||
M24157.1 | - | 2 | 192 | |||
M28129.1 | - | 2 | 196 | |||
M30510.1 | - | 2 | 177 | |||
X55988.1 | - | 2 | 218 |