Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005977.3 | NP_005968.1 | 3 | 483 | ||
NM_183043.2 | NP_898864.1 | 3 | 224 | |||
NM_183044.2 | NP_898865.1 | 3 | 358 | |||
NM_183045.1 | NP_898866.1 | 3 | 483 | |||
XM_005266485.2 | XP_005266542.1 | 3 | 829 | |||
XM_005266486.2 | XP_005266543.1 | 3 | 277 | |||
XM_011535177.2 | XP_011533479.1 | 3 | 851 | |||
XM_011535178.1 | XP_011533480.1 | 3 | 882 | |||
XM_017020685.1 | XP_016876174.1 | 3 | 882 | |||
XM_017020686.1 | XP_016876175.1 | 3 | 433 | |||
XM_017020687.1 | XP_016876176.1 | 3 | 277 | |||
GenBank mRNA | AF293342.1 | - | 3 | 373 | ||
AJ010346.1 | - | 3 | 373 | |||
AJ010347.1 | - | 3 | 174 | |||
AK293272.1 | - | 3 | 145 | |||
AK312435.1 | - | 3 | 428 | |||
AY009109.1 | - | 3 | 306 | |||
BC034688.2 | - | 2 | 124 | 4508997 | ||
BI461269.1 | - | 3 | 488 | |||
BP310130.1 | - | 3 | 297 | |||
CR749238.1 | - | 3 | 415 | |||
DB062601.1 | - | 3 | 360 | |||
DC310579.1 | - | 3 | 224 | |||
KU178309.1 | - | 1 | 13 | |||
KU178310.1 | - | 1 | 13 |