Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000991.4 | NP_000982.2 | 3 | 136 | ||
NM_001136134.1 | NP_001129606.1 | 3 | 136 | |||
NM_001136135.1 | NP_001129607.1 | 3 | 136 | |||
NM_001136136.1 | NP_001129608.1 | 3 | 136 | |||
NM_001136137.1 | NP_001129609.1 | 3 | 136 | |||
GenBank mRNA | AB209627.1 | - | 2 | 119 | ||
AK096870.1 | - | 3 | 1618 | |||
AK293736.1 | - | 3 | 134 | |||
AK311760.1 | - | 3 | 136 | |||
BC010173.2 | - | 3 | 121 | 4054251 | ||
BC010182.2 | - | 3 | 125 | 4301420 | ||
BC011582.1 | - | 2 | 115 | 4561118 | ||
BP227047.1 | - | 3 | 136 | |||
CR457024.1 | - | 2 | 94 | |||
DA092122.1 | - | 3 | 136 | |||
DA333143.1 | - | 3 | 135 | |||
DA820669.1 | - | 3 | 136 | |||
U14969.1 | - | 2 | 121 |