Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135771.2 | NP_001129243.1 | 10, 11 | 1520 | ||
NM_001324299.1 | NP_001311228.1 | 11, 12 | 1616 | |||
NM_001324301.1 | NP_001311230.1 | 12, 13 | 1664 | |||
NM_001324302.1 | NP_001311231.1 | 11, 12 | 1616 | |||
NM_001324303.1 | NP_001311232.1 | 11, 12 | 1616 | |||
NM_001324304.1 | NP_001311233.1 | 11, 12 | 1616 | |||
NM_001324305.1 | NP_001311234.1 | 12, 13 | 1664 | |||
NM_001324306.1 | NP_001311235.1 | 9, 10 | 1344 | |||
NM_002951.4 | NP_002942.2 | 11, 12 | 1616 | |||
XM_006723851.2 | XP_006723914.1 | 12, 13 | 1469 | |||
XM_006723852.2 | XP_006723915.1 | 12, 13 | 1469 | |||
GenBank mRNA | AK096243.1 | - | 10, 11 | 1405 | ||
AK296126.1 | - | 9, 10 | 1019 | |||
AK308683.1 | - | 9, 10 | 1043 | |||
AK315801.1 | - | 11, 12 | 1344 | |||
BC002380.2 | - | 8, 9 | 890 | |||
BC003560.1 | - | 11, 12 | 1342 | 3546673 | ||
BC013028.2 | - | 3, 4 | 256 | |||
BC020222.1 | - | 11, 12 | 1487 | 3640766 | ||
CR456899.1 | - | 11, 12 | 1305 | |||
Y00282.1 | - | 11, 12 | 1593 |