Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324299.1 | NP_001311228.1 | 3, 4 | 607 | ||
NM_001324301.1 | NP_001311230.1 | 3, 4 | 607 | |||
NM_001324302.1 | NP_001311231.1 | 3, 4 | 607 | |||
NM_001324303.1 | NP_001311232.1 | 3, 4 | 607 | |||
NM_001324304.1 | NP_001311233.1 | 3, 4 | 607 | |||
NM_001324305.1 | NP_001311234.1 | 3, 4 | 607 | |||
NM_002951.4 | NP_002942.2 | 3, 4 | 607 | |||
XM_006723851.2 | XP_006723914.1 | 3, 4 | 412 | |||
XM_006723852.2 | XP_006723915.1 | 3, 4 | 412 | |||
GenBank mRNA | AK315801.1 | - | 3, 4 | 335 | ||
BC003560.1 | - | 3, 4 | 333 | 3546673 | ||
BC020222.1 | - | 3, 4 | 478 | 3640766 | ||
BG718075.1 | - | 3, 4 | 612 | |||
CR456899.1 | - | 3, 4 | 296 | |||
Y00282.1 | - | 3, 4 | 584 |