Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SORT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205228.1 | NP_001192157.1 | 4, 5 | 564 | ||
NM_002959.6 | NP_002950.3 | 4, 5 | 592 | |||
XM_005271100.2 | XP_005271157.1 | 4, 5 | 587 | |||
XM_005271101.2 | XP_005271158.1 | 4, 5 | 706 | |||
XM_005271102.1 | XP_005271159.1 | 4, 5 | 626 | |||
XM_006710812.1 | XP_006710875.1 | 4, 5 | 362 | |||
GenBank mRNA | AK293118.1 | - | 4, 5 | 550 | ||
AK301548.1 | - | 4, 5 | 567 | |||
AK310147.1 | - | 4, 5 | 455 | |||
BC023542.1 | - | 4, 5 | 588 | 4123836 | ||
DB045034.1 | - | 4, 5 | 304 |